Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 12.18 mm Diameter, Tool Steel
Supplier:  MSC 68103233
Encompass
This Class ZZ Plus Plug Gage is designed for "Go Inspection" and for setting micrometers, verniers, and other measuring devices. It has a diameter of 12.18 mm, an overall length of 2.0000 inches, and is made of durable tool steel. The gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Class ZZ Plus plug gage
  • 12.18 mm diameter
  • 2.0000 in overall length
  • Tolerance of 0.00508 mm
  • 10 Micro finish
  • Constructed from tool steel
  • Ideal for "Go Inspection" applications
  • Useful for setting micrometers, verniers, and other measuring devices
  • Supplied with a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
Catalog No. 50-005-31348