Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 15.91 mm, Tool Steel
Supplier: MSC 68106962
This Vermont Gage Class ZZ Plus plug gage is designed for precise "Go" inspection and for setting micrometers, verniers, and other measuring devices. It features a 15.91 mm diameter with a 0.00508 mm tolerance and a 10 micro finish, ensuring high accuracy in measurement applications. Made from durable tool steel, it offers reliability and extended service life.
- Includes certificate of accuracy with NIST traceability.
- Suitable for "Go" inspection.
- Ideal for setting micrometers, verniers, and other measuring devices.
- Constructed from durable tool steel.
- Features a 10 micro finish for precision.
- Meets ASME B89.1 standards.