Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 3.80 mm, 2.0000 in
Supplier: MSC 68094853
This plug gage is a Class ZZ Plus model, featuring a 3.80 mm diameter and an overall length of 2.0000 inches. It is constructed from tool steel and is suitable for go inspection, as well as for setting micrometers, verniers, and other measuring devices. Each gage includes a certificate of accuracy with NIST traceability.
- Constructed from tool steel
- Designed for go inspection
- Can be used to set micrometers, verniers, and other measuring devices
- Includes a certificate of accuracy with NIST traceability
- Meets ASME B89.1 standards
- Features a 10 micro finish
- Tolerance of 0.00508 mm