Learn More
MSC Vermont Gage Plug & Pin Gage, 20.55 mm Diameter, Class ZZ Minus, Steel
Supplier: MSC 40448003
This Class ZZ Minus Plug & Pin Gage features a 20.55 mm diameter with a precise 0.00508 mm tolerance. Constructed from steel, it is designed for accurate measurement and inspection tasks and has an overall length of 2 inches.
- Includes certificate of accuracy with NIST traceability
- Coating/finish: 10 micro
- Suitable for NoGo inspection
- Can be used to set micrometers, verniers, and other measuring devices