Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1140 in Dia, 2 in OAL
Supplier:  MSC 89011407
Encompass
This plug and pin gage is a Class ZZ Minus gage with a 0.1140 inch diameter and 2 inch overall length. It is designed for applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices. The gage is constructed from steel and features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from steel.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-30223