Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1300 in Diameter, 2 in Overall Length
Supplier:  MSC 36871598
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. With a 0.1300 inch diameter and a 2 inch overall length, it is suitable for tasks such as setting micrometers, performing NoGo inspection, and calibrating verniers and other measuring devices.
  • Features a black oxide finish
  • Constructed from durable steel material
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, and other measuring devices
Catalog No. 50-005-28339