Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1565 in, 2 in OAL
Supplier:  MSC 89015655
Encompass
The Class ZZ Minus Plug & Pin Gage features a diameter of 0.1565 inches and an overall length of 2 inches. It offers a tolerance of 0.000200 inches and is crafted from steel with a 10 Micro coating/finish. This gage is designed for applications such as setting micrometers, performing NoGo inspection, and use with verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-31374