Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.3020 Inch Diameter, Steel
Supplier: MSC 08417354
This Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection and setting of various measuring devices like micrometers and verniers. Manufactured from durable steel with a black oxide finish, it ensures reliable performance and resistance to wear. Each gage comes with a Certificate of Accuracy, traceable to NIST standards, guaranteeing its precision and quality.
- Precision ground to Class ZZ minus tolerance.
- Ideal for setting micrometers, verniers, and for no-go inspection.
- Constructed from high-quality steel for durability.
- Features a black oxide coating for enhanced wear resistance.
- Includes a certificate of accuracy with NIST traceability.
- Provides reliable and accurate measurements for quality control.
- Overall length of 2 inches.