Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.8350 in Dia, 2 in OAL, Steel
Supplier: MSC 89083505
This product is a Class ZZ Minus Plug & Pin Gage with a 0.8350-inch diameter and a 2-inch overall length. It is made of steel and features a 10 Micro finish. It is designed for various measuring applications, including setting micrometers, NoGo inspection, and use with verniers and other measuring devices. A Certificate of Accuracy with NIST Traceability is included.
- Class ZZ Minus gage type.
- 0.8350 in diameter.
- 2 in overall length.
- Steel material.
- 0.000200 in tolerance.
- 10 Micro finish.
- Includes certificate of accuracy with NIST traceability.
- Suitable for setting micrometers.
- Suitable for NoGo inspection.
- Suitable for verniers and other measuring devices.