Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.9290 in Diameter, 2 in Overall Length, Steel
Supplier:  MSC 89092902
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. It features a 0.9290 inch diameter and an overall length of 2 inches, constructed from durable steel.
  • Measures 0.9290 inch in diameter
  • Has an overall length of 2 inches
  • Features a ZZ Minus class for precise fits
  • Offers a tight tolerance of 0.000200 inch
  • Constructed from steel for durability
  • Includes a 10 micro coating/finish
  • Supplied with a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, no-go inspection, verniers, and other measuring devices
  • Manufactured in the United States
Catalog No. 50-005-31092