Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 15.40 mm Diameter, 2 in OAL
Supplier: MSC 40442857
This plug and pin gage is a Class ZZ Minus tool, designed for precise measurements. It features a 15.40 mm diameter and a 2 inch overall length, making it suitable for various inspection and calibration tasks. Crafted from durable steel with a 10 Micro coating, it is built for accuracy and includes a certificate of accuracy with NIST traceability.
- Class ZZ Minus
- 15.40 mm diameter of measurement
- 2 inch overall length
- 0.00508 mm tolerance
- Steel material
- 10 Micro coating/finish
- Application for setting micrometers, NoGo inspection, verniers, and other measuring devices
- Includes a certificate of accuracy with NIST traceability
- Manufactured in the United States
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.