Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 5.42 mm, 2 in OAL
Supplier:  MSC 40432874
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for various gaging applications. It features a 5.42 mm diameter and a 2 in overall length. Constructed from durable tool steel with a 10 microinch finish, it is ideal for setting micrometers, NoGo inspection, and checking hole sizes.
  • Class ZZ tolerance for precise measurements
  • Minus tolerance direction for even or whole sizes
  • 5.42 mm diameter
  • 2 in overall length
  • Made from durable tool steel
  • Features a 10 microinch finish or better
  • Includes a certificate of accuracy for quality assurance
  • NIST traceable for reliable standards
  • Ideal for Go/NoGo gaging, measuring hole sizes, and setting micrometers
Catalog No. 50-005-27029