Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 7.43 mm Diameter, 2 inch Length
Supplier:  MSC 40434888
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurately setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. It offers high precision and comes with a Certificate of Accuracy with NIST traceability.
  • Precision measuring tool
  • Designed for setting micrometers, NoGo inspection, and calibrating measuring devices
  • Constructed from steel for durability
  • Features a 10 micro finish
  • Includes a certificate of accuracy with NIST traceability
  • 7.43 mm diameter
  • 2 inch overall length
Catalog No. 50-005-28897