Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 8.69 mm, 10 Microfinish
Supplier:  MSC 40436149
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool with an 8.69 mm diameter, designed for no-go inspection. It features a tolerance of 0.00508 mm.
  • Steel material
  • Application for no-go inspection
  • Includes certificate of accuracy with NIST traceability
  • 10 microfinish coating
  • 2 in overall length
  • 0.00508 mm tolerance
  • 8.69 mm diameter of measurement
Catalog No. 50-005-28469