Learn More
MSC Vermont Gage Threaded Ring Gage, #3-48 UNC, Class 2A, No Go
Supplier:  MSC 55322697
Encompass
This threaded ring gage is designed for precise measurement of #3-48 UNC threads. It is a Class 2A "No Go" gage, ensuring accurate verification of thread tolerance. Constructed from durable tool steel, it meets NIST standards for quality and reliability.
  • Designed for #3-48 UNC threads.
  • Features Class 2A tolerance for accuracy.
  • Functions as a "No Go" gage.
  • Made from durable tool steel.
  • Meets NIST standards.
  • Single ring design.
  • Manufactured in the United States.
Catalog No. 50-005-30269