Learn More
MSC Vermont Gage Threaded Ring Gage, #4-48 UNF, Class 2A, Go
Supplier:  MSC 05414487
Encompass
The product is a Threaded Ring Gage designed for inspecting threads. It is specifically a "Go" gage, meaning it checks if a manufactured thread is not too large and will allow the mating part to assemble. It is made of Tool Steel and adheres to NIST standards.
  • Go gage
  • #4-48 thread size
  • UNF thread type
  • 2A classification
  • Single ring
  • Tool steel material
  • NIST standards compliant
  • Made in United States
Catalog No. 50-005-29399