Learn More
MSC Vermont Gage Threaded Ring Gage, 7/8-14 UNF, Class 2A, No Go, Tool Steel
Supplier:  MSC 05428156
Encompass
This threaded ring gage is designed for precise measurement of external threads. It features a 7/8-14 UNF thread size and a Class 2A classification, ensuring accurate gaging for quality control applications. Constructed from durable tool steel, this "No Go" gage helps verify that threads are within specified tolerances, preventing oversized threads.
  • Designed for measuring external threads.
  • Features a 7/8-14 UNF thread size.
  • Class 2A classification for accuracy.
  • No Go configuration to prevent oversized threads.
  • Constructed from high-quality tool steel for durability.
  • Meets NIST standards.
Catalog No. 50-005-29400