Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 0.9245 in., Class ZZ, 1 ea
Supplier: MSC 06940282
This Class ZZ steel pin gage is designed for precise measurement applications, providing a reliable standard for checking hole sizes, hole locations, and distance between holes. It features a plus tolerance and is manufactured for consistent accuracy in machining and inspection environments.
- Constructed from durable tool steel
- Hardness rating of 60/65 Rc
- Finish of 10 microinch or better
- Reversible style design
- Diameter and direction of tolerance marked on the gage
- Certificate of accuracy included