Learn More
Medchemexpress LLC Vernakalant (hydrochloride) | 748810-28-8 | MFCD09833303 | 99.3% | C20H32ClNO4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1418350MG
Encompass_Preferred
Vernakalant hydrochloride is a small molecule drug approved in 2010 for atrial fibrillation. It functions as a mixed voltage- and frequency-dependent Na+ and atria-preferred K+ channel blocker, targeting specific Kv1.5 channels.
  • Potassium channel blocker
  • Mixed voltage- and frequency-dependent Na+ channel blocker
  • Soluble in DMSO and water
  • Stable under recommended storage conditions
  • Solid form, white to light yellow appearance
Catalog No. 50-003-53482