Learn More
ATCC Vero Kidney African Green Monkey (Chlorocebus aethiops) Grivet
Supplier:  ATCC CCL81
Encompass
The Vero cell line was initiated in 1962 from the kidney tissue derived from a normal, adult African green monkey. The cell line can be used in a variety of applications, including the detection of verotoxins, detection of virus in ground beef, efficacy testing, the study of malaria, media testing, and mycoplasma testing.
Catalog No. 50-238-2893