Learn More
ATCC Vero-SF-ACF; Kidney; African Green Monkey (Chlorocebus aethiops) Grivet
Supplier:  ATCC CCL81.5
Encompass
Vero-SF-ACF is a cell line exhibiting epithelial morphology that was adapted to serum-free and animal component-free medium. This cell line can be used in the detection of verotoxin, efficacy testing, malaria biology, media testing, and mycoplasma testing.
Catalog No. 50-238-4790