Learn More
Medchemexpress LLC Verrucarin A | 456069 | MFCD00056759 | 99.9% | 502.55 g/mol | C27H34O9 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1074261MG
Encompass_Preferred
Verrucarin A is a Type D macrocyclic mycotoxin derived from Myrothecium verrucaria that acts as a potent inhibitor of protein synthesis. It is supplied as a small, high-purity research reagent for in vitro studies of apoptosis, cell signaling, and cell-cycle regulation.
  • High purity suitable for biochemical and cell-based assays.
  • Small pack size ideal for scarce or potent compounds.
  • Inhibits protein synthesis and induces apoptosis in cell models.
  • Useful for studying MAPK/ERK/Akt signaling and cell-cycle effects.
  • Handled as a highly toxic compound with appropriate safety precautions.
Catalog No. 50-002-40553