Learn More
Foxx Life Sciences EZBio GL45 Cap with VersaCap
Supplier:  Foxx Life Sciences 2051003RLS
Encompass_Preferred
Foxx Life Sciences’ EZBio GL45 Cap with VersaCap technology is leak-proof and provides the ultimate in top-connection flexibility for plastic and glass bottles. Open VersaCap is compatible with a variety of adapter inserts, enabling users to quickly change the top connections of each bottle to match their application needs. VersaCap offers finger grips that allow for easy tightening and opening. The EZBio GL45 Cap includes molded-in hose barb (HB) options, available in a closed cap, open cap and closed adapter, 2x 1⁄8” HB, 2x 1⁄4”HB, 3x 1⁄8” HB, 4x 1⁄8” HB in both white and natural colors. All EZBio GL45 Caps and adapters are made from polypropylene, which may be autoclaved at 15 PSI and 121°C (250°F) for 20 minutes.
Catalog No. NC1516338