Learn More
MSC Clorox Healthcare VersaSure Cleaner Disinfectant Wipes, Refill, 1-Ply, 12 in x 10 in
Supplier:  MSC 65556045
Encompass
Clorox Healthcare VersaSure Cleaner Disinfectant Wipes are alcohol-free quat wipes designed for broad-spectrum disinfection. These refill wipes are versatile, killing bacteria, viruses, and fungi in 2 minutes or less. They offer a low-residue, one-step cleaning and disinfecting solution.
  • Alcohol-free quat wipes provide broad-spectrum disinfection.
  • Kill bacteria, viruses, and fungi in 2 minutes or less.
  • One-step solution for cleaning and disinfecting surfaces.
  • Low residue and low odor formula.
  • Designed for compatibility with common healthcare surfaces.
  • Patented technology enhances quat activity for faster kill times.
Catalog No. 50-283-7609