Learn More
Accuflow Systems Saint-Gobain Versilon Reinforced TPU Tubing, 3/4 in ID, 30 m
Supplier:  Accuflow Systems AZY02054
Encompass_Preferred
Versilon C-544-A IB braid-reinforced TPU tubing provides abrasion-resistant, high-pressure fluid transfer for process and instrumentation applications. The ether-based polyurethane formulation retains flexibility at low temperatures and resists oils, greases, and abrasion, while braid reinforcement enables tight-bend performance and elevated working pressures.
  • Reinforced ether-based polyurethane (TPU) construction for elevated working pressures.
  • 3/4 in inner diameter (19.05 mm), 1-1/16 in outer diameter (26.99 mm), 5/32 in wall thickness (3.97 mm).
  • Clear color for visual fluid monitoring.
  • 30 m (98.4 ft) coil length for continuous runs.
  • Resistant to abrasion, oils, greases, and tearing.
  • Suitable for process lines, instrumentation, slurry transfer, and coolant recovery.
Catalog No. NC3244597