Learn More
Cayman Chemical VerteporfIn 10mg
SDP
Supplier:  Cayman Chemical 1733410 MG
Encompass_Preferred
A photosensitizer used during photodynamic therapy to eliminate abnormal blood vessels in the eye that are associated with conditions such as macular degeneration; also inhibits autophagosome formation by directly targeting and modifying p62, a scaffold and adaptor protein that binds both polyubiquitinated proteins destined for degradation and LC3 on autophagosomal membranes
Catalog No. 50-297-9647