Learn More
Apexbio Technology LLC Verteporfin, 129497-78-5, MFCD11982858, 5 mg
SDP
Supplier:  Apexbio Technology LLC A83275
Encompass_Preferred
MF: C41H42N4O8, MW: 718.79, Synonyms: CL 318952; Visudyne. Solubility: 18.3mg/mL in DMSO. Verteporfin, derived from porphyrin, is a potent photosensitizing agent, which has all the features theoretically necessary for effective photodynamic therapy (PDT) in ocular neovascularization.
Catalog No. 50-101-3744