Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Half (Slim) Rack for 3" boxes, Holds 5 boxes
SDP
Supplier:  Alkali Scientific LR53
Encompass
For Standard 3″ Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 3 inch boxes. These slim style vertical freezer racks allow you to maximize your freezers capacity storage by filling in gaps that can’t fit standard chest/LN2 freezer racks. Each slim style vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimension H x D x W: 26 3/4 x 3 1/4 x 5 9/16 inch (679 x 82 x 141mm)Compartment Size H x D x W: 5 5/16 x 3 1/4 x 5 1/4 inch (134.5 x 82 x 133mm)  
Catalog No. 50-302-8816