Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for 25-Place Slide Boxes, Holds 6 boxes
SDP
Supplier:  Alkali Scientific CFS256
Encompass
For 25-Place Slide Boxes The CFS-25-6 vertical freezer racks for storage of 25-place slide boxes, features a 6 boxes high configuration for storage in a laboratory chest or LN2 freezer. These vertical freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer and also features locking rods for added security. This vertical freezer rack does not come standard with 25-place slide boxes. Dimensions H x D x W: 9 7/16 x 3 13/16 x 6 inch (239 x 97 x 152mm)Compartment Dimensions h x d x w: 1 1/2 x 3 11/16 x 5 5/8 inch ( 38 x 93 x 143mm) *Security lock device is ordered separately.
Catalog No. 50-302-9344