Learn More
Labrepco Inc VERTICAL RACKS FOR CHEST FRZ
SDP
Supplier:  Labrepco Inc CF9281
Encompass_Preferred
Vertical Racks for Chest Freezer and Liquid NitrogenTank for Standard2" BoxesRack w/ Cardboard Box and Divider (81cell)19 7/8" x 5 5/8" x 5 1/2" or 505 x 143 x 140mmShipping Weight (lbs.): 6Capacity: 9
Catalog No. NC3232654