Learn More
Enzo Life Sciences Vesicular stomatitis virus (VSV-G) tag polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADIMSA115E
Encompass_Preferred
Host: Rabbit. Immunogen: Vesicular stomatitis virus (VSV-G) tag (YTDIEMNRLGK) coupled to GST. Species reactivity: Human, Mouse, Rat. Applications: ICC, WB. Recommended Dilutions: Immunocytochemistry (10µg/ml) Western Blot (1:100, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7348