Learn More
Perkin Elmer US LLC Vespel Ferrule, 0.0625 in
Supplier:  Perkin Elmer US LLC 04974018
Encompass_Preferred
Vespel ferrules are not reusable and not recommended for use with fused silica capillary columns. Quantity: Package of 10 Fitting Size: 1/8 inch Ferrule Inner Diameter: 1/8 inch
Catalog No. 50-211-1907