Learn More
Perkin Elmer US LLC Vespel Ferrule, 1.6 mm I.D.
Supplier:  Perkin Elmer US LLC 04972263
Encompass_Preferred
For use with 1/16 inch metal tubing and glass-lined recievers. Temperature limit is 350 °C.
Catalog No. 50-211-3300