Learn More
Fine Science Tools VESSEL CANNULATION FORCEPS11.5
SDP
Supplier:  Fine Science Tools 1840311
Encompass
Product: Vessel Cannulation Forceps - 0.35mm OD/11.5cm/ Tip Shape: Angled/ Tubing Size: 0.35mm OD/ Alloy: Inox/ Length: 11.5cm/ Unit: Each
Catalog No. NC1109025