Learn More
World Precision Instrument Vessel Clip, Straight, 0.75 x 4mm Jaw, German
SDP
Supplier:  World Precision Instrument 501779G
Encompass
Micro Vessel Clips are a surgical instrument for use in microvascular procedures, where extremely small blood vessels and delicate tissues are involved. WPI Micro Vessel Clips are significantly smaller than standard surgical clips, because they are designed to occlude fine blood vessels with precise control. These clips are made of German stainless steel and come in a couple of sizes. These ligating vessel clips can tie off blood vessels or other tissues. Micro vessel clips are made to be durable and withstand the stresses of surgery, ensuring that they remain securely in place throughout the procedure. Pinch the handle to release pressure and position the vessel clip, or release the handles to clamp the tissue or vessel. The size of the vessel clip you need can vary depending on the size of the vessel or tissue being occluded. These are available with 0.8x6mm, and 0.75x4mm jaws.
Catalog No. 50-822-824