Learn More
Med Vet International Ivermectin Injection, Vet use, 1%, 50ml
Supplier:  Med Vet International PHIVOMEC
Encompass_Preferred
Large Animal NOT FOR SALE IN CALIFORNIAIvermax (ivermectin) Injection is an injectable parasiticide for cattle and swine. One low-volume dose effectively treats and controls the following internal and external parasites that may impair the health of cattle and swine: gastrointestinal roundworms (including inhibited Ostertagia ostertagi in cattle), lungworms, grubs, sucking lice, and mange mites of cattle; and gastrointestinal roundworms, lungworms, lice, and mange mites of swine. Ivermectin’s convenience, broad-spectrum efficacy and safety margin make Ivermax Injection a unique product for parasite control of cattle and swine.Products are not always manufacturer specific. Actual product appearance may differ from website image.
Catalog No. 50-304-4897