Learn More
Medchemexpress LLC HY-137458A 5mg , Vevorisertib (trihydrochloride) CAS:1416775-08-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY137458A5MG
Encompass_Preferred
HY-137458A 5mg Vevorisertib (trihydrochloride) CAS: 1416775-08-0 Vevorisertib (ARQ 751) trihydrochloride is a selective, allosteric, pan-AKT and AKT1-E17K mutant inhibitors. Vevorisertib trihydrochloride potently inhibit phosphorylation of AKT. Vevorisertib trihydrochloride has Kd values of 1.2 nM and 8.6 nM for AKT1 and AKT1-E17K, respectively. Vevorisertib trihydrochloride has IC50 values of 0.55, 0.81, and 1.3 nM for AKT1, AKT2, and AKT3, respectively. Vevorisertib trihydrochloride can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1416