Learn More
Cayman Chemical VH 032 amIde C7 AcId 50mg
SDP
Supplier:  Cayman Chemical 4044750 MG
Encompass_Preferred
A PROTAC precursor; has been used in the synthesis of PROTACs targeting ALK in cancer cells
Catalog No. 50-294-3078