Learn More
Cayman Chemical VH 032 LInker 1 25mg
SDP
Supplier:  Cayman Chemical 2614825 MG
Encompass_Preferred
A PROTAC precursor; has been used in the synthesis of PROTACs targeting TEAD in cancer cells
Catalog No. 50-293-5066