Learn More
Cayman Chemical VH 032 LInker 2 5mg
SDP
Supplier:  Cayman Chemical 261495 MG
Encompass_Preferred
A VHL Ligand 1 derivative; comprises the VHL ligand domain and PEG linker of the BET inhibitor-containing PROTACs MZ2 and MZP-55
Catalog No. 50-293-3925