Learn More
Medchemexpress LLC VH 032, phenol | 2244684-42-0 | MFCD34368523 | 99.4% | C24H32N4O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13616410MG
Encompass_Preferred
VH032-OH is a VH032-derived von Hippel-Lindau (VHL) E3 ligase ligand for research use. It serves as a synthetic building block for PROTAC design, providing a phenolic handle for linker attachment and conjugation to target-binding ligands.
  • High purity (99.4%) suitable for analytical and synthetic research.
  • Phenolic functional group for versatile linker conjugation.
  • Suitable as an E3 ligase recruiting element in PROTAC assembly.
  • Well-defined molecular properties (C24H32N4O5S; MW 488.60) for planning syntheses.
  • Available in small-mass quantities for rapid screening and method development.
Catalog No. 50-002-59211