Learn More
Medchemexpress LLC VH 101 phenol-alkylC4-amine dihydrochloride | 2564467-03-2 | >95.0% | 676.67 g/mol | C30H44Cl2FN5O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1393355MG
Encompass_Preferred
VH 101 phenol-alkylC4-amine dihydrochloride is a functionalized von Hippel-Lindau (VHL) E3 ligase ligand supplied as the dihydrochloride salt. It is designed as a building block for PROTAC (proteolysis-targeting chimera) synthesis and degrader design, offering a linker-equipped ligand with a terminal amine for conjugation and medicinal chemistry applications.
  • Functionalized VHL ligand for PROTAC synthesis.
  • Dihydrochloride salt for improved stability and handling.
  • Terminal amine compatible with common coupling chemistries.
  • Alkyl C4 linker suitable for assembling bifunctional degraders.
  • Available in small milligram quantities for medicinal chemistry research.
Catalog No. 50-002-23436