Learn More
Medchemexpress LLC HY-100947 5mg Medchemexpress, VH-298 CAS:2097381-85-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1009475MG
Encompass_Preferred
Medchemexpress, HY-100947 5mg VH-298 CAS:2097381-85-4 VH-298 is a highly potent inhibitor of the VHL:HIF-α interaction with a K d value of 80 to 90 nM, used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1997