Learn More
Medchemexpress LLC VH032-C7-COOH | 2172819-77-9 | 99.9% | 614.80 g/mol | C32H46N4O6S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1360055MG
Encompass_Preferred
VH032-C7-COOH is a functionalized von Hippel-Lindau (VHL) protein ligand developed for PROTAC research and conjugation chemistry. It incorporates an E3 ligase-binding motif attached through an alkyl linker terminating in a carboxylic acid, enabling coupling to target ligands. The compound is provided at high purity and is intended for research applications in targeted protein degradation and medicinal chemistry.
Catalog No. 50-002-22548