Learn More
Selleck Chemical LLC (S.R.S)-AHPC-PEG4-NH2 hydrochloride S3502-5mg
SDP
Supplier:  Selleck Chemical LLC S35025MG
Encompass_Preferred
(S R S)-AHPC-PEG4-NH2 hydrochloride (VH032-PEG4-NH2 hydrochloride VHL Ligand-Linker Conjugates 4 hydrochloride E3 ligase Ligand-Linker Conjugates 7) is a PROTAC E3 ligase ligand-linker conjugate that incorporates the (S R S)-AHPC based VHL ligand and 4-unit PEG linker
Catalog No. 50-310-9842