Learn More
Medchemexpress LLC Vh285-peg4-c4-cl | 1799506-07-2 | MFCD34601688 | >98.0% | 799.42 g·mol⁻¹ | C41H55ClN4O8S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1119975MG
Encompass_Preferred
VH285-PEG4-C4-Cl is a VHL-ligand-HaloTag conjugate designed as a targeted protein degrader for use in cellular and biochemical assays. It connects a VH285-based VHL ligand to a HaloTag-targeting moiety via a PEG4-C4 linker to induce proteasomal degradation of HaloTag fusion proteins. Supplied for research use with high stated purity and available in small quantities or as a DMSO solution.
  • Highly potent degrader with a DC50 of 19 nM.
  • Can induce up to 90% degradation of GFP-HaloTag7 in cells.
  • VHL ligand-HaloTag conjugate with a PEG4-C4 linker for targeted engagement.
  • Supplied with reported purity ≥98% and molecular weight 799.42 g·mol⁻¹.
  • Available in small mg quantities or as a 10 mM solution in DMSO for reconstitution.
  • Suitable for cellular and biochemical studies of targeted protein degradation.
Catalog No. 50-002-63807