Learn More
Medchemexpress LLC VH285-PEG4-C4-Cl (HaloPROTAC 3) | 1799506-07-2 | ≥98.0% | 799.42 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11199750MG
Encompass_Preferred
VH285-PEG4-C4-Cl (HaloPROTAC 3) is a conjugate of ligands for E3 and a 16-atom-length linker. This degrader incorporates the VH285 based VHL ligand and an alkyl/ether-based linker.
  • Highly potent and efficacious degrader of GFP-HaloTag7 with a DC50 of 19 nM.
  • Able to induce 90% degradation of GFP-Halotag at 625 nM.
  • Binds to VHL with an IC50 of 0.54 μM.
Catalog No. 50-004-39354