Learn More
Medchemexpress LLC VHL protein, human (His) | >90.0% | 5 UG

Supplier: Medchemexpress LLC HYP761245UG
Recombinant human von Hippel-Lindau (VHL) protein with an N-terminal His tag, expressed in Escherichia coli and supplied lyophilized for biochemical and assay applications. The product spans amino acids M1-D213 (≈34 kDa) and is provided at high purity for binding studies, structural analysis, and assay development.
- source organism: human
- expression system: E. coli
- tag: N-6xHis
- sequence range: M1-D213
- molecular weight: ≈34 kDa
- purity: >90% by reducing SDS-PAGE
- formulation: lyophilized with 6% trehalose in Tris-HCl or PBS
- reconstitution: do not reconstitute below 100 μg/mL
- storage: store at -20°C; aliquot and store at -80°C for long term
- available size: includes 5 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.