Learn More
Beckman Coulter Vi-CELL XR Concentration Control
Supplier:  Beckman Coulter 175478
Encompass_Preferred
For use with the Vi-CELL XR Cell Viability Analyzer. The concentration control consists of a mixture of latex beads at 1x106 beads/mL concentration. It is recommended to run daily to verify that the Vi-CELL is counting properly. Item must be refrigerated after receiving.
Catalog No. NC9552505