Learn More
Polysciences, Inc. ViaCheck Viability Control 90%
SDP
Supplier:  Polysciences, Inc. 2462520
Encompass
Image based instrument viability controls. Our particle standards offer discrete "live" and "dead" populations. They can be delivered in customizable ratios & concentrations. Count: 1 x 106. Storage: Do not permit to freezeViaCheck Viability Control 90% is a mixed suspension of microspheres that simulates a 90:10 ratio of live and dead (or dying) cells stained with Trypan Blue.
Catalog No. NC9678338